1-Brom-4-heptylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: p-Brom-heptylbenzol

OS-7610-25G OS-7610-1G OS-7610-5G
COBBOS-7610-25GEA 227 EUR
COBBOS-7610-25G COBBOS-7610-1G COBBOS-7610-5G
1-Brom-4-heptylbenzol
1-Brom-4-heptylbenzol
Formel: C₁₃H₁₉Br
Molecular Weight: 255,2 g/mol
MDL: MFCD00061115
CAS-Nummer: 76287-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR