4-Methoxybenzophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
p-Methoxybenzophenon
, 4-Benzoylanisol
M-Clarity™ quality level = MQ200
SIALM10301-25GEA
27.1
EUR
SIALM10301-25G
4-Methoxybenzophenon, Sigma-Aldrich®
4-Methoxybenzophenon
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 354 °C (1013 hPa) Schmelzpunkt: 59…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008403 CAS-Nummer: 611-94-9 EINECS: 210-285-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...