2-Amino-3-brom-5-methylpyridin

Lieferant: COMBI-BLOCKS INC.
Achtung

Synonyme: 3-Brom-5-methyl-2-pyridylamin2-Amino-3-brom-5-picolin3-Brom-5-methylpyridin-2-amin

COBBOT-0343-5GEA 25.1 EUR
COBBOT-0343-5G COBBOT-0343-25G COBBOT-0343-100G
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR