2-Amino-5-methylthiazol

Lieferant: COMBI-BLOCKS INC.

Synonyme: 5-Methylthiazol-2-ylamin

COBBHI-1203-25GEA 47.9 EUR
COBBHI-1203-25G COBBHI-1203-100G
2-Amino-5-methylthiazol
2-Amino-5-methylthiazole
Formel: C₄H₆N₂S
Molecular Weight: 114,17 g/mol
Siedepunkt: 222 °C (1013 hPa)
Schmelzpunkt: 96 °C
MDL: MFCD00078317
CAS-Nummer: 7305-71-7
EINECS: 423-800-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR