2-Fluorbenzolsulfonamid

Lieferant: FLUOROCHEM LTD (CHEMPUR)

Synonyme: 2-Fluorbenzolsulfonsäureamid

CHMPCFF264.5EA 348 EUR
CHMPCFF264.5 FLUO017311-1G FLUO017311-5G CHMPCFF264.2.5
2-Fluorbenzolsulfonamid
2-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
Molecular Weight: 175,18 g/mol
MDL: MFCD00042283
CAS-Nummer: 30058-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR