Naphthol AS-D-chloracetat
Lieferant: Molekula
Synonyme: 3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthylchloracetat
MOLE31848932-1GEA 252 EUR
MOLE31848932-1G
Naphthol AS-D-chloracetat
Naphthol AS-D-chloracetat
Formel: C₂₀H₁₆ClNO₃
Molecular Weight: 353,8 g/mol
CAS-Nummer: 35245-26-2
EINECS: 252-463-5