3-Brom-2-chlor-5-methylpyridin

Lieferant: COMBI-BLOCKS INC.

Synonyme: 3-Brom-2-chlor-5-picolin

COBBOT-0345-5GEA 36.9 EUR
COBBOT-0345-5G COBBOT-0345-100G COBBOT-0345-1G COBBOT-0345-25G
3-Brom-2-chlor-5-methylpyridin
3-Bromo-2-chloro-5-methylpyridine
Formel: C₆H₅BrClN
MDL: MFCD01830664
CAS-Nummer: 17282-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR