4-Brom-2-methylbenzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
4-Brom-2-methylbenzol-1-sulfonylchlorid, 4-Brom-2-methylbenzolsulfochlorid, 4-Brom-2-methylbenzolsulfonsäurechlorid
RDP00246.0001
RDP00246DA
RDP00246EA
MAYBRDP00246.0001EA
0
EUR
MAYBRDP00246.0001
MAYBRDP00246DA
MAYBRDP00246EA
4-Brom-2-methylbenzolsulfonylchlorid
4-Brom-2-methylbenzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,55 g/mol |
MDL:
MFCD00219927 CAS-Nummer: 139937-37-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...