4-Methyl-3-nitrobenzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 4-Methyl-3-nitrobenzol-1-sulfonylchlorid4-Methyl-3-nitrobenzolsulfochlorid4-Methyl-3-nitrobenzolsulfonsäurechlorid

MAYBKM08627.50EA 0 EUR
MAYBKM08627.50
4-Methyl-3-nitrobenzolsulfonylchlorid
4-Methyl-3-nitrobenzolsulfonylchlorid
Formel: C₇H₆ClNO₄S
Molecular Weight: 235,65 g/mol
MDL: MFCD00129811
CAS-Nummer: 616-83-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR