Digitoxin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3β,14β-Dihydroxy-5β-carden-20(22)-olid-3-tridigitoxid
M-Clarity™ Qualitätsstufe = MQ200
SIALD5878-500MGEA
186
EUR
SIALD5878-500MG
SIALD5878-250MG
SIALD5878-5G
SIALD5878-1G
SIALD5878-10G
Digitoxin, Sigma-Aldrich®
Digitoxin
Formel:
C₄₁H₆₄O₁₃ Molecular Weight: 764,95 g/mol Schmelzpunkt: 276 °C |
MDL:
MFCD00003686 CAS-Nummer: 71-63-6 EINECS: 200-760-5 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...