Palladium(II)acetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PalladiumdiacetatEssigsäure Palladium(II)salz

M-Clarity™ Qualitätsstufe = MQ200

SIAL205869-250GEA 0 EUR
SIAL205869-250G SIAL205869-2G SIAL205869-10G SIAL205869-1G SIAL205869-1KG
Palladium(II)acetat, Sigma-Aldrich®
Palladium(II)acetat
Formel: C₄H₆O₄Pd
Molecular Weight: 224,51 g/mol
Schmelzpunkt: 205 °C
MDL: MFCD00012453
CAS-Nummer: 3375-31-3
EINECS: 222-164-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR