Octachlornaphthalin

Lieferant: EHRENSTORFER GMBH
Achtung

EHERC20425800EA 49 EUR
EHERC20425800
Octachlornaphthalin
Octachlornaphthalin
Formel: C₁₀Cl₈
Molecular Weight: 403,73 g/mol
CAS-Nummer: 2234-13-1
EINECS: 218-778-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR