n-Dotetracontan, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

44250-100MGEA 91.5 EUR
44250-100MG
n-Dotetracontan, Supelco®
n-Dotetracontane
Formel: H₃C(CH₂)₄₀CH₃
Molecular Weight: 591,14 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7098-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR