n-Heptan, EMPLURA®, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
n-Dipropylmethan
M-Clarity™ Qualitätsstufe = MQ200
1.04365.9011EA
387
EUR
1.04365.9011
1.04365.2511
1.04365.2500
1.04365.6025
1.04365.6190
1.04365.9950
1.04365.1000
n-Heptan, EMPLURA®, Supelco®
n-Heptan
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel:
H₃C(CH₂)₅CH₃ Molecular Weight: 100,20 g/mol Siedepunkt: 98,4 °C (1013 hPa) Schmelzpunkt: –90,6 °C Dichte: 0,6838 g/cm³ (20 °C) Flammpunkt: –4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009544 CAS-Nummer: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II Merck Index: 13,04679 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...