n-Hexan, EMPLURA®, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Hexyl hydride, Hexanen, Hexane
M-Clarity™ Qualitätsstufe = MQ200
1.04368.1000 1.04368.2500 1.04368.6025 1.04368.6190 1.04368.9190 1.04368.2511 1.04368.9011
1.04368.1000EA 62.1 EUR
1.04368.1000 1.04368.2500 1.04368.6025 1.04368.6190 1.04368.9190 1.04368.2511 1.04368.9011
n-Hexan, EMPLURA®, Supelco®
n-Hexan
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel: H₃C(CH₂)₄CH₃
Molecular Weight: 86,18 g/mol
Siedepunkt: 69 °C (1013 hPa)
Schmelzpunkt: –94,3 °C
Dichte: 0,659 g/cm³ (20 °C)
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM