n-Nonadecan
Lieferant: Molekula
MOLEM67720907EA
0
EUR
MOLEM67720907
n-Nonadecan
n-Nonadecan
Formel:
H₃C(CH₂)₁₇CH₃ Molecular Weight: 268,53 g/mol Siedepunkt: 329,7 °C (1013 hPa) Schmelzpunkt: 32,5 °C Dichte: 0,7845 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009012 CAS-Nummer: 629-92-5 EINECS: 211-116-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...