n-Tetracontan ≥97%
Lieferant: Thermo Scientific
11274.02EA
77.3
EUR
11274.02
11274.04
n-Tetracontan ≥97%
n-Tetracontane
Purity: 97% min
CAS No.: 4181-95-7
Molecular Formula: C40H82
Formula Weight: 563.09
Physical Form: Flakes
Density: 0.779
Appearance: White
Melting Point: 80-84°
Boiling Point: 150°/0.001mm
Beilstein Reference: 1800152
MDL No.: MFCD00015267
Formel:
H₃C(CH₂)₃₈CH₃ Molecular Weight: 563,09 g/mol Siedepunkt: 150 °C (0,001 mmHg) Schmelzpunkt: 80…84 °C Dichte: 0,779 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015267 CAS-Nummer: 4181-95-7 EINECS: 224-055-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...