n-Tetracontan, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
87086-250MGEA
0
EUR
87086-250MG
n-Tetracontan, Supelco®
n-Tetracontane
Formel:
H₃C(CH₂)₃₈CH₃ Molecular Weight: 563,09 g/mol Schmelzpunkt: 82…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015267 CAS-Nummer: 4181-95-7 EINECS: 224-055-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...