n-Tetracosan ≥99%

Lieferant: Thermo Scientific

42462.06EA 24.1 EUR
42462.06 42462.22 42462.14
n-Tetracosan ≥99%
n-Tetracosane

Purity: 99%
CAS No.: 646-31-1
Molecular Formula: CH3(CH2)22CH3
Formula Weight: 338.66
Flash Point: 110°(230°F)
Physical Form: Crystalline
Melting Point: 49-52°
Boiling Point: 391°
MDL No.: MFCD00009352

Formel: H₃C(CH₂)₂₂CH₃
Molecular Weight: 338,66 g/mol
Siedepunkt: 391 °C
Schmelzpunkt: 49…52 °C
Dichte: 0,7787 g/cm³ (20 °C)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009352
CAS-Nummer: 646-31-1
EINECS: 211-474-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR