DL-Cycloserin

Lieferant: BACHEM BIOCHEMICA

Synonyme: DL-4-Aminoisoxazolidin-3-on(±)-Cycloserin

BACE4005653.0050EA 150 EUR
BACE4005653.0050 BACE4005653.0250 BACE4005653.1000
DL-Cycloserin
DL-Cycloserin
Formel: C₃H₆N₂O₂
Molecular Weight: 102,09 g/mol
MDL: MFCD00064323
CAS-Nummer: 68-39-3
EINECS: 200-687-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR