4-Nitrophenylhydrazin

Lieferant: Thermo Scientific

MAYBBTB09059.10EA 0 EUR
MAYBBTB09059.10 MAYBBTB09059.50 MAYBBTB09059.1
4-Nitrophenylhydrazin
4-Nitrophenylhydrazin
Formel: (C₆H₄(NO₂))NHNH₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 210…212 °C
CAS-Nummer: 100-16-3
EINECS: 202-824-8
UN: 3376
ADR: 4.1,I
Merck Index: 12,06721

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR