Pentachloranisol

Lieferant: EHRENSTORFER GMBH

Synonyme: 2,3,4,5,6-Pentachloranisol

EHERC15950000EA 84 EUR
EHERC15950000
Pentachloranisol
Pentachloranisol
Formel: C₇H₃Cl₅O
Molecular Weight: 280,36 g/mol
CAS-Nummer: 1825-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR