2,4,6-Triallyloxy-1,3,5-triazin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,4,6-Tris(allyloxy)-s-triazin, 2,4,6-Triallyloxy-s-triazin, 2,4,6-Tris(allyloxy)-1,3,5-triazin, Cyanursäuretriallylester
M-Clarity™ quality level = MQ100
291609-100G
291609-500G
SIAL291609-100GEA
81.5
EUR
SIAL291609-100G
SIAL291609-500G
2,4,6-Triallyloxy-1,3,5-triazin, Sigma-Aldrich®
2,4,6-Triallyloxy-1,3,5-triazin
Formel:
C₁₂H₁₅N₃O₃ Molecular Weight: 249,27 g/mol |
MDL:
MFCD00006049 CAS-Nummer: 101-37-1 EINECS: 202-936-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...