DL-Cycloserin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DL-4-Aminoisoxazolidin-3-on(±)-Cycloserin

M-Clarity™ quality level = MQ200

SIALC7005-250MGEA 992 EUR
SIALC7005-250MG
DL-Cycloserin, Sigma-Aldrich®
DL-Cycloserin
Formel: C₃H₆N₂O₂
Molecular Weight: 102,09 g/mol
MDL: MFCD00064323
CAS-Nummer: 68-39-3
EINECS: 200-687-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR