(±)-β-Citronellol, Supelco®
Lieferant: Merck
Synonyme:
(±)-Citronellol
51381-1MLEA
314
EUR
51381-1ML
(±)-β-Citronellol, Supelco®
(±)-β-Citronellol
Formel:
C₁₀H₂₀O Molecular Weight: 156,27 g/mol Siedepunkt: 222 °C (1013 hPa) Dichte: 0,86 g/cm³ (20 °C) Flammpunkt: 100 °C |
MDL:
MFCD00002935 CAS-Nummer: 106-22-9 EINECS: 203-375-0 UN: 3082 ADR: 9,III Merck Index: 13,02354 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...