3-Brom-o-toluidin Hydrochlorid
Lieferant: COMBI-BLOCKS INC.
COBBHC-6940-5GEA
291
EUR
COBBHC-6940-5G
COBBHC-6940-1G
3-Brom-o-toluidin Hydrochlorid
3-Brom-o-toluidin Hydrochlorid
Formel:
C₇H₉BrClN |
MDL:
MFCD10699676 CAS-Nummer: 16822-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...