o-Dianisidin Dihydrochlorid ≥95%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 3,3'-Dimethoxybenzidine dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

D3252-100G D3252-25G D3252-5G
D3252-100GEA 1220 EUR
D3252-100G D3252-25G D3252-5G
o-Dianisidin Dihydrochlorid ≥95%, Sigma-Aldrich®
o-Dianisidin Dihydrochlorid

o-Dianisidine (3,3′-dimethoxybenzidine) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is yellow-orange in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 5 M HCl.


  • Physical form: Crystalline


o-Dianisidine (3,3′-dimethoxybenzidine) has been used as a component of GO (glucose oxidase) reagent, which is used for measuring the glucose content. It has also been used as a substrate to measure myeloperoxidase activity.


Bitte beachten Probably carcinogenic.

Formel: C₁₄H₁₈Cl₂N₂O₂
Molecular Weight: 317,21 g/mol
Schmelzpunkt: 268 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012488
CAS-Nummer: 20325-40-0
EINECS: 243-737-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27db5b6542ac074f22f94a0233419856a69083d4&vid=b83a250167ad03154b8f2f1caac63a9121246746&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Light Purple and Faint Red to Light Red and Faint Beige to Light Beige and Faint Grey to Light Grey and Pink
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Turbidity)
25 mg/ml, H₂O
Clear to Very Slightly Hazy
Solubility (Colour) Colorless to Faint Yellow and Colorless to Light Red
Water (by Karl Fischer)
Carbon (anhydrous) 50.1 - 55.7%
Nitrogen (anhydrous) 8.3 - 9.3%
Titration by AgNO₃ (anhydrous)
% C
21.1 - 23.5%
Recommended Retest Period 4 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR