Methyl-4-indolcarboxylat 99%

Lieferant: Thermo Scientific
Achtung

ACRO334030010EA 49.1 EUR
ACRO334030010
Methyl-4-indolcarboxylat 99%
Methyl-4-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 67…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191222
CAS-Nummer: 39830-66-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to green Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 71 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR