5,8,11,14-Eicosatetrainsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: ETYAOctadehydroarachidonic acid

M-Clarity™ quality level = MQ200

SIALE1768-5MGEA 293 EUR
SIALE1768-5MG
5,8,11,14-Eicosatetrainsäure, Sigma-Aldrich®
5,8,11,14-Eicosatetraynoic acid
Formel: C₂₀H₂₄O₂
MDL: MFCD00036967
CAS-Nummer: 1191-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR