5,8,11,14-Eicosatetrainsäure, Millipore®
Lieferant: Merck Millipore Calbiochem
434741-20EA 420 EUR
434741-20
5,8,11,14-Eicosatetrainsäure, Millipore®
5,8,11,14-Eicosatetraynoic acid
Formel: C₂₀H₂₄O₂
Molecular Weight: 296,41 g/mol
MDL: MFCD00036967
CAS-Nummer: 1191-85-1