β-Jonon, Sigma-Aldrich®
Lieferant: Merck
|
|
M-Clarity™ Qualitätsstufe = MQ200
8.18619.0100EA
84.8
EUR
8.18619.0100
8.18619.9999
β-Jonon, Sigma-Aldrich®
β-Jonon
Formel:
C₁₃H₂₀O Molecular Weight: 192,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001549 CAS-Nummer: 79-77-6 EINECS: 201-224-3 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | -49 °C |
Molar Mass | 192.3 g/mol |
Boiling Point | 127 - 128 °C (13 hPa) |
Flash Point | 122 °C |
Refractive Index | 1.5192 (20 °C, 589 nm) |
Density | 0.94 g/cm3 (20 °C) |
Hill Formula | C13H20O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...