4'-Cyanbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO020754-1GEA 918 EUR
FLUO020754-1G
4'-Cyanbiphenyl-4-carbonsäure
4'-Cyanbiphenyl-4-carbonsäure
Formel: C₁₄H₉NO₂
Molecular Weight: 223,23 g/mol
MDL: MFCD00438676
CAS-Nummer: 5728-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR