4-Aminobenzamidoxim 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Aminobenzamide oxime, 4-Amino-N'-hydroxybenzimidamide

H51788.03 H51788.06
H51788.03EA 75 EUR
H51788.03 H51788.06
4-Aminobenzamidoxim 97%
4-Aminobenzamidoxime

Purity: 97% CAS No.: 277319-62-7 Molecular Formula: C7H9N3O Formula Weight: 151.17 Melting Point: 168-170° MDL No.: MFCD08061151

Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08061151
CAS-Nummer: 277319-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR