4-Pentyloxyanilin

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO005867-5GEA 0 EUR
FLUO005867-5G FLUO005867-25G
4-Pentyloxyanilin
4-Pentyloxyanilin
Formel: C₁₁H₁₇NO
MDL: MFCD00007867
CAS-Nummer: 39905-50-5
EINECS: 254-695-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR