Lidocain, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALL7757-25GEA 71.4 EUR
SIALL7757-25G SIALL7757-100G
Lidocain, Sigma-Aldrich®
Lidocain
Formel: C₁₄H₂₂N₂O
MDL: MFCD00026733
CAS-Nummer: 137-58-6
EINECS: 205-302-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR