Mehrstellen-Magnetrührer mit Heizplatte, AM4
Lieferant: VELP Scientifica
442-1113EA 1530 EUR
442-1113
Mehrstellen-Magnetrührer mit Heizplatte, AM4
Rührer Mehrstellen-Magnetrührer mit Heizung
Multiple-position hot plate stirrer with four separately controlled stirring plates coated with aluminium alloy which provides uniform heat distribution.
  • AM4 has four independently controlled positions
  • High power PCM-type driving magnet
  • Control panel is IP 42 protected from liquid spills with run-off groove
IP protection class according to DIN EN 60529: IP 21.