p-Xylol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,4-Dimethylbenzol

A10534.AEEA 25.7 EUR
A10534.AE A10534.0F A10534.AP
p-Xylol ≥99%
p-Xylol

Purity: 99%
CAS No.: 106-42-3
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 27°(80°F)
Refractive Index: 1.4950
Odour: Aromatic
Melting Point: 12-13°
Boiling Point: 137-138°
Beilstein Reference: 1901563
Merck Reference: 14,10081
UN No.: 1307
MDL No.: MFCD00008556

Formel: C₆H₄(CH₃)₂
Molecular Weight: 106,17 g/mol
Siedepunkt: 137…138 °C
Schmelzpunkt: 12…13 °C
Dichte: 0,866
Flammpunkt: 27 °C (80 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008556
CAS-Nummer: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 14,10081
https://de.vwr.com/stibo/web/std.lang.all/87/75/768775.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR