4-(Benzyloxy)benzaldehyd
Lieferant: FLUOROCHEM LTD (CHEMPUR)
FLUO018216-10GEA
0
EUR
FLUO018216-10G
4-(Benzyloxy)benzaldehyd
4-(Benzyloxy)benzaldehyd
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 366 °C (1013 hPa) Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003387 CAS-Nummer: 4397-53-9 EINECS: 224-527-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...