6-Chinolylmethylether ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A11736.14EA
109
EUR
A11736.14
A11736.06
A11736.22
6-Chinolylmethylether ≥98%
6-Chinolylmethylether
Purity: 98%
CAS No.: 5263-87-6
Molecular Formula: C10H9NO
Formula Weight: 159.19
Flash Point:
Refractive Index: 1.6250
Melting Point: 17-20°
Boiling Point: 141-145°/15mm
Beilstein Reference: 115244
MDL No.: MFCD00006800
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Siedepunkt: 141…145 °C (15 mmHg) Schmelzpunkt: 17…20 °C Dichte: 1,152 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006800 CAS-Nummer: 5263-87-6 EINECS: 226-077-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...