Saccharin Natrium

Lieferant: CAESAR & LORENTZ GMBH

CSLO2578.0100EA 30.3 EUR
CSLO2578.0100
Saccharin Natrium
Saccharin Natrium
Formel: C₇H₅NNaO₃S
Molecular Weight: 206,18 g/mol
MDL: MFCD00013092
CAS-Nummer: 128-44-9
EINECS: 204-886-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR