Calciumbis(dihydrogenorthophosphat) Monohydrat
Lieferant: VWR Chemicals
22403.262EA
0
EUR
22403.262
Calciumbis(dihydrogenorthophosphat) Monohydrat
Calciumbis(dihydrogenorthophosphat) Monohydrat
Formel:
H₄CaO₈P₂·1H₂O Molecular Weight: 252,07 g/mol |
MDL:
MFCD00149602 CAS-Nummer: 10031-30-8 EINECS: 231-837-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...