Brij® 35, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Brij® L23
M-Clarity™ Qualitätsstufe = MQ200
P1254-500G
P1254-5KG
SIALP1254-500GEA
76.7
EUR
SIALP1254-500G
SIALP1254-5KG
Brij® 35, Sigma-Aldrich®
Brij® 35
Formel:
C₅₈H₁₁₈O₂₄ Molecular Weight: 1199,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080891 CAS-Nummer: 9002-92-0 EINECS: 500-002-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...