Brij® 35

Lieferant: Uptima
Achtung

Synonyme: Brij® L23

UPTIUP091871EA 0 EUR
UPTIUP091871 UPTIUP091870
Brij® 35
Brij® 35
Formel: C₅₈H₁₁₈O₂₄
Molecular Weight: 1199,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00080891
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR