HONC60240

Lieferant: Honeywell Chemicals
Gefahr

60240-1KGEA 132.02 EUR
60240-1KG HONC60240-25KG 60240-2.5KG HONC60240-10KG 60240-250G
HONC60240
Potassium fluoride

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: KF
Molecular Weight: 58,1 g/mol
Siedepunkt: 1,505 °C (1.013 hPa)
Schmelzpunkt: 846 °C
Dichte: 2,48 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011398
CAS-Nummer: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 12,07794

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 1000 ppm
Chloride (Cl) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR