Natriumiodid ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Iodnatrium
11665.22
11665.36
11665.A3
11665.22EA
72.4
EUR
11665.22
11665.36
11665.A3
Natriumiodid ≥99,9% (Metall-Basis)
Natriumiodid
Purity: 99.9%
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: Powder
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 651°
Boiling Point: 1300-1304°
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formel:
NaI Molecular Weight: 149,89 g/mol Siedepunkt: 1300…1304 °C Schmelzpunkt: 651 °C Dichte: 3,67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003532 CAS-Nummer: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III Merck Index: 14,8631 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...