Natriumsulfit

Lieferant: EMD MILLIPORE CORPORATION

EMSCSX0785-20EA 0 EUR
EMSCSX0785-20
Natriumsulfit
Sodium sulphite
Formel: Na₂SO₃
Molecular Weight: 126,04 g/mol
Schmelzpunkt: > 500 °C
Dichte: 2,63 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003503
CAS-Nummer: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR