Benzylharnstoff ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
B24945.22EA
87
EUR
B24945.22
B24945.14
Benzylharnstoff ≥98%
Benzylharnstoff
Formel:
C₈H₁₀N₂O Molecular Weight: 150,18 g/mol Schmelzpunkt: 146…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007951 CAS-Nummer: 538-32-9 EINECS: 208-689-1 Merck Index: 14,1147 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...