Methyl-5-bromnicotinat
Lieferant: Thermo Scientific
Synonyme: Methyl 5-bromonicotinate
RF04619DA RF04619EA
MAYBRF04619DAEA 0 EUR
MAYBRF04619DA MAYBRF04619EA
Methyl-5-bromnicotinat
Methyl-5-bromnicotinat
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD00173668
CAS-Nummer: 29681-44-5