2-Brom-3-chlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-3-chloropyridine-4-boronic acid

BB-8565-5G BB-8565-1G
COBBBB-8565-5GEA 473 EUR
COBBBB-8565-5G COBBBB-8565-1G
2-Brom-3-chlor-4-pyridinboronsäure
2-Bromo-3-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
MDL: MFCD08690231
CAS-Nummer: 1003043-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR