2-Bromisonicotinsäure
Lieferant: COMBI-BLOCKS INC.
COBBCA-4041-25GEA
60.8
EUR
COBBCA-4041-25G
COBBCA-4041-100G
COBBCA-4041-5G
2-Bromisonicotinsäure
2-Bromisonicotinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202,01 g/mol |
MDL:
MFCD01646069 CAS-Nummer: 66572-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...